HTL 5145 Optipette OP12-10 Multichannel Pipettor, Red, 12-Channel; 0.5 to 10 µL
Representative image only.
HTL 5145 Optipette OP12-10 Multichannel Pipettor, Red, 12-Channel; 0.5 to 10 µL
| Mfr: 5145| Item: 2455175
Easily adjust ejector height
Call For Price
Call For Price
Product Description
Key Features
  • Ergonomic handle design
  • Easy to calibrate
  • Rotating pipetting push button prevents accidental volume change
  • 3 year warranty
More About this Item
Remarkably robust and durable in a sleek design, Optipette adjustable-volume pipettors offer optimal accuracy and precision. With a UV-resistant body and autoclavable bottom half, the soft spring system reduces pipetting forces while color-coded rings simplify volume identification. Additional spacers enable ejector height adjustment; user-calibration is easy thanks to a provided calibration key.
3 Year Warranty
Specifications
  • AutoclavableYes
  • Max Volume (µL)10
  • Min Volume (µL)0.5
  • Accuracy (%)±2
  • Precision (%)±1.2
  • Number Of Channels12
  • Tip EjectorYes
  • Color CodeRed
  • DescriptionMultichannel Pipettor, Red, 12-Channel; 0.5 to 10 µL