HTL 5144 Optipette OP8-300 Multichannel Pipettor, Green, 8-Channel; 50 to 300 µL
Representative image only.
HTL 5144 Optipette OP8-300 Multichannel Pipettor, Green, 8-Channel; 50 to 300 µL
| Mfr: 5144| Item: 2455174
Easily adjust ejector height
Call For Price
Call For Price
Product Description
Key Features
  • Ergonomic handle design
  • Easy to calibrate
  • Rotating pipetting push button prevents accidental volume change
  • 3 year warranty
More About this Item
Remarkably robust and durable in a sleek design, Optipette adjustable-volume pipettors offer optimal accuracy and precision. With a UV-resistant body and autoclavable bottom half, the soft spring system reduces pipetting forces while color-coded rings simplify volume identification. Additional spacers enable ejector height adjustment; user-calibration is easy thanks to a provided calibration key.
3 Year Warranty
Specifications
  • AutoclavableYes
  • Max Volume (µL)300
  • Min Volume (µL)50
  • Accuracy (%)±1
  • Precision (%)±0.6
  • Number Of Channels8
  • Tip EjectorYes
  • Color CodeGreen
  • DescriptionMultichannel Pipettor, Green, 8-Channel; 50 to 300 µL