Horiba 3200156563 Multiparameter Meter, 6-ft (2-m) Cable

Representative image only.

Horiba 3200156563 Multiparameter Meter, 6-ft (2-m) Cable

Horiba | Mfr: 3200156563| Item: 9960025

Measure and display pH, mV, conductivity, TDS, salinity, dissolved oxygen, turbidity, seawater specific gravity, water depth, and temperature simultaneously

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Horiba 3200156563 Multiparameter Meter, 6-ft (2-M) Cable
Horiba 3200156563 Multiparameter Meter, 6-ft (2-M) Cable
9960025

Call For Price
+
Horiba 362176 Replacement Combination PH Electrode
9959050

₹38,100.00INR / Each
+ =
Total Price 62800.00 ADD 3 ITEMS TO CART
Product Description

Includes

one probe, one 500-mL bottle of pH 4.0 buffer solution, one 250-mL bottle of KCl solution, DO sensor installation set, cleaning brush, calibration cup, batteries, shoulder strap, and carrying case.

Key Features

  • Auto-calibration feature simplifies calibration of pH, dissolved oxygen, conductivity, turbidity, and depth
  • Built-in GPS — ideal for environmental surveys
  • Rugged housing is both waterproof and shock resistant
  • Stores up to 10,000 readings with USB connectivity for export of data to a PC
  • 1 year warranty

More About this Item

These handheld, waterproof meters feature a large 3-1/2" (8.9 cm), easy-to-read LCD with backlighting for use in dark and low-light conditions. Multiple parameters can be measured and displayed at the same time. USB/probe connectivity, GPS info, and battery power are also indicated via the display. Variable contrast allows you to adjust for extreme ambient light conditions.

Meters are designed for one-handed operation. Probes are attached via a quick-connect connector allowing for fast setup. Unique auto-calibration feature provides hassle-free meter setup. Turbidity sensor offers better accuracy than most other conventional water quality meters. Polarographic DO sensor features improved stability, fast response, and easy maintenance.

Auto-hold function freezes average data values providing more time to verify or transcribe data. Up to 10,000 data sets can be stored and later transferred to a PC via a built-in USB connection. Units of measure are user selectable which helps you save time.

1 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier
Specifications
  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.1
  • Buffer RecognitionDIN, NIST, and USA
  • Min Conductivity0 mS
  • Max Conductivity100 mS
  • Conductivity Resolution0.001, 0.01, or 0.1 mS; 0.001 or 0.01 S
  • Conductivity Accuracy±1% FS
  • Turbidity Resolution0.1 NTU
  • Dissolved Oxygen Resolution0.01 ppm
  • Temperature compensationAutomatic
  • InterfaceUSB
  • Data Logging (points)10,000 data sets
  • Width (cm)11.43
  • Height (cm)28.2575
  • Width (in)4 1/2
  • Length (in)2 5/8
  • Height (in)11 1/8
  • DescriptionMultiparameter Meter, 6-ft (2-m) Cable
  • Warranty1 year
Reviews
Q&A