Honeywell DR Graphic Recorder with TrendServer Pro Software

Representative image only.

Honeywell DR Graphic Recorder with TrendServer Pro Software

Honeywell | Mfr: TVDRG2-040-12-0-010-00000S-000| Item: 4903107

Go paperless!

Call For Price

for an additional
₹40,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Honeywell DR Graphic Recorder with TrendServer Pro Software
Honeywell DR Graphic Recorder with TrendServer Pro Software
4903107

Call For Price
+ =
Total Price 40300.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Identical panel cutout to Honeywell DR4300 and DR4500 aids the transition away from chart recording
  • Chart-paper-like display is familiar and easy to read
  • Touch screen, data storage, and communications features make programming simple and expand capabilities
  • 18 month warranty

More About this Item

This paperless recorder lets you upgrade from traditional chart paper recording to paperless recording without requiring a new panel build if you have the Honeywell DR4300 or DR4500 recorder currently. The display mimics a circular chart, so it is visually similar, but without the inconvenience and potential downtime associated with pens and chart paper.


Use the supplied configuration credits to customize your recorder. Each model is programmed with ten credits to use in any way. All functions are programmed in the firmware, but must be unlocked using the credits. You can mix and match and reprogram at any time. Available options include maintenance, printing, extra digital pens, configuration lockout, remote viewing, reports, e-mail, secure communications, custom screens, maths, and much more.


Choose your software:
TrendManager Pro software offers all of the functions of TrendViewer and adds full recorder configuration and data export capabilities.

TrendServer Pro software builds on the features of TrendManager Pro by adding network connectivity with FTP and web browser access and real-time data acquisition.


Many more input/output, power, memory, and credit configurations are available through special order.

18 Month Warranty
Specifications
  • Number of Channels4
  • Temperature AccuracyThermocouple: ±1.2°F (0.7°C); RTD: ±0.5°F (0.3°C)
  • Output TypeEthernet, USB (host function for external devices or flash drives), Modbus® (optional with credits)
  • Memory Size1 GB solid state flash; external USB flash drive or mass storage device (up to 120 GB, not included) and SD card (up to 32 GB, not included)
  • Sampling Rate1 second
  • Display TypeXGA touch-screen TFT LCD, 12-1/8" (30.7 cm) high
  • AlarmAlarm set points: 6 per pen, configurable to change screen background color and trigger alarm relay; Alarm relay contact: SPNC rated at 1 A, 24 V
  • Width (in)14 7/8
  • Height (in)15 1/4
  • Depth (in)5 1/8
  • Width (cm)38
  • Height (cm)38.7
  • Depth (cm)13
  • Power (VAC)100 to 240
  • DescriptionGraphic Recorder with TrendServer Pro Software
  • Warranty18 months
Reviews
Q&A