Honeywell DL5PPB-300-00 Dissolved Oxygen Probe, PVC Body
Representative image only.
Honeywell DL5PPB-300-00 Dissolved Oxygen Probe, PVC Body
Honeywell | Mfr: DL5PPB-300-0000-000| Item: 5661140
Based on equilibrium technology—offers excellent accuracy
₹730,000.00INR / Each
₹730,000.00INR / Each
Product Description
Key Features
  • No need to recalibrate after cleaning
  • Does not require membrane replacement
  • 20 ft (6.1 m) long cable for remote installation
  • 1" NPT(M) process connection for immersion applications
  • 1 year warranty
More About this Item
Keep your oxygen meter running properly with dissolved oxygen probes.
Honeywell dissolved oxygen (DO) electrode features a PVC body with a 20 ft (6.1 m) long cable and an 1" NPT(M) process connection for immersion applications.
1 Year Warranty
Specifications
  • MaterialPVC
  • Compatible WithHoneywell Multi-Input Controller
  • Connections1" NPT(M)
  • DescriptionDissolved Oxygen Probe, PVC Body