Honeywell 8Y3032/9Q North Hypalon drybox gloves, 30 mil, size 9-3/4, 1 pair
Representative image only.
Honeywell 8Y3032/9Q North Hypalon drybox gloves, 30 mil, size 9-3/4, 1 pair
Honeywell | Mfr: 8Y3032/9Q| Item: 8656912
Shoulder-length, seamless gloves for critical working conditions
₹222,000.00INR / Pair of 1
₹222,000.00INR / Pair of 1
Product Description
More About this Item
Hypalon® gloves offer outstanding abrasion protection and resistance to ozone or oxidizing chemicals. Natural softness improves worker comfort.
Specifications
  • MaterialHypalon®
  • Thickness (mil)30
  • SizeMedium
  • Length (in)32
  • Length (cm)81
  • ColorWhite
  • ComplianceANSI/ISEA 105 - ASTM D3389-05 Abrasion Test, ANSI/ISEA 105 - ASTM D412 - Physical properties, ANSI/ISEA 105 - ASTM F1790-97 Cut Test, EN374 - Permiation and breakthrough time, EN388 - Mechanical risks from abrasion, cut, tear and puncture, EN420 - General Requirements, OSHA Standard -29 CFR 1915.157
  • SterileNo
  • DescriptionHypalon drybox gloves, 30 mil, size 9-3/4, 1 pair