Honeywell 50016134-501 Software, program and analyze, w/ FTP and web browser
Representative image only.
Honeywell 50016134-501 Software, program and analyze, w/ FTP and web browser
Honeywell | Mfr: 50016134-501| Item: 8066057
Cost-effective electronic data collection in a compact DIN-sized recorder
₹456,000.00INR / Each
for an additional
₹40,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹456,000.00INR / Each
Buy It Together
Current Item
Honeywell 50016134-501 Software, program and analyze, w/ FTP and web browser
Honeywell 50016134-501 Software, program and analyze, w/ FTP and web browser
8066057
₹456,000.00INR / Each
+ +
Honeywell 50016133-501 Software, program and analyze
8066059
₹282,000.00INR / Each
=
Total Price 778300.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Program up to 6 user-definable soft alarms per pen to announce out-of-limit conditions
  • Durable touch screen provides easy data entry and rapid navigation through menus
  • 18 month warranty
More About this Item
These electronic recorders provide flexible, general-purpose data recording that enables you to monitor and control your process instrumentation with ease. Logging rates and chart speeds can be programmed independently allowing data to be displayed and stored at the best-suited rate. All data logged is automatically time-and-date stamped with a real time clock. These recorders provide enhanced security options that protect programs from accidental change with allowing up to four levels of password protection and 10 different passwords.
The large, full-color display shows charts, digital values, and trends for quick and easy operator interpretation. The intuitive touch screen menus make set up and navigation simple, and provide easy-to-read data allowing the operator to make quick decision
18 Month Warranty
Specifications
  • Power (VAC)115 / 230
  • DescriptionSoftware, program and analyze, w/ FTP and web browser
  • Warranty18 months