Honeywell 4214 Salisbury Insulated Universal Switch Stick, 6 foot length, 1.25" diameter
Representative image only.
Honeywell 4214 Salisbury Insulated Universal Switch Stick, 6 foot length, 1.25" diameter
Honeywell | Mfr: 4214-ES| Item: 8643708
₹188,000.00INR / Each
₹188,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Also know as hotsticks, these insulated switch sticks are used for opening and closing disconnect switches and cutouts. Also used for installing and removing open-link fuse links. Sticks have a 1.25" (32 mm) diameter and a prong head.
1 Year Warranty
Specifications
  • Length (InchShort)72
  • Diameter (InchShort)1 1/4
  • DescriptionInsulated Universal Switch Stick, 6 foot length, 1.25" diameter