Honeywell 32-000513-0000 pH balanced, bacteria-inhibiting saline eyewash solution
Representative image only.
Honeywell 32-000513-0000 pH balanced, bacteria-inhibiting saline eyewash solution
Honeywell | Mfr: 320005130000H5| Item: 0679664
NOT AVAILABLE IN CANADA
Call For Price
Call For Price
Product Description
Key Features
  • Solution improves the safety and performance of self-contained eyewash stations
More About this Item
Mix one 180-oz bottle of Eyesaline® solution (06796-64) with water to fill your 14- to 16-gallon self-contained eyewash station. Eyesaline solution is pH balanced to the human eye to offer preserved, buffered relief and prevent the likelihood of additional eye damage.
Solution has a two-year shelf life and is stamped with lot number and date code.
Specifications
  • DescriptionpH balanced, bacteria-inhibiting saline eyewash solution