Honeywell 20817 Salisbury Insulated Static Discharge Stick, 4-ft L, 6-ft Grounding Cable
Representative image only.
Honeywell 20817 Salisbury Insulated Static Discharge Stick, 4-ft L, 6-ft Grounding Cable
Honeywell | Mfr: 20817| Item: 8643701
Remove static charge after de-energizing a circuit to improve worker safety
Call For Price
Call For Price
Product Description
Key Features
  • 1 year warranty
More About this Item
Safely remove static charge after de-energizing with these static discharge sticks. Stick comes preassembled with a brass alloy U-hook and is constructed with closed cell foam-filled tubular fiberglass, made in accordance with ASTM F711. Includes 6-ft (1.8 m) of copper grounding cable attached to a flat jaw grade 3 bronze serrated clamp with strain relief.
1 Year Warranty
Specifications
  • DescriptionInsulated Static Discharge Stick, 4-ft L, 6-ft Grounding Cable