Honeywell 12" Classic Circular Chart Recorder, Single Pen/Channel with Display
Representative image only.
Honeywell 12" Classic Circular Chart Recorder, Single Pen/Channel with Display
Honeywell | Mfr: DR45A1-1000-00-000-0-000000-0| Item: 8066111
Combines the simplicity of pen drawn analog traces with the sophistication of microprocessor controlled functions
₹1,240,000.00INR / Each
₹1,240,000.00INR / Each
Buy It Together
Current Item
Honeywell 12" Classic Circular Chart Recorder, Single Pen/Channel with Display
Honeywell 12" Classic Circular Chart Recorder, Single Pen/Channel with Display
8066111
₹1,240,000.00INR / Each
+ =
Total Price 1250500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Preprinted charts meet specific recording needs
  • User configurable operating parameters can be set using the clear display and tactile keypad
  • Disposable ink cartridge for reliable recording with minimal maintenance
  • Digital PID controller lets users configure exact control action
  • Plug and play tuning algorithms with Accutune II™
  • 18 month warranty
More About this Item
Aimed at applications in food processing, pharmaceuticals, heat treatment, environmental testing, manufactured goods, and water treatment, this recorder eliminates the need for a stock of preprinted charts - it creates its own, providing flexibility to users. It is a microprocessor-based recorder suitable to meet a wide range of industry requirements. It comes with a digital display and easy-to-use operator interface. The Truline pen provides a convenient method to document a process while minimizing maintenance hassles. An integral PID controller and RS-485 Modbus communications ensure user empowerment. This accurate and easily configurable recorder is designed to work in tough industrial situations.
Specifications
  • Number of Channels1
  • Chart TypeCircular
  • Chart Speed8 hrs, 12 hrs, 24 hrs, 7 days, or selected hours per revolution
  • Chart Diameter (in)12
  • Chart Width (in)4.62
  • Pen Response Time6-seconds maximum with no filtering
  • Tip StyleDisposable fiber-tip
  • Pen ColorPurple
  • Input Type+20 Vdc source for external dry contact or isolated solid state contacts. Selects one configured input.
  • Sampling RateEach input sampled 3 times a second
  • Display TypeVacuum fluorescent, alphanumeric
  • AlarmYes
  • Description12" Classic Circular Chart Recorder, Single Pen/Channel with Display
  • Warranty18 months