Hirschmann 9392050 Solarus Electronic Burette, Bottletop Dispenser; 50 mL

Representative image only.

Hirschmann 9392050 Solarus Electronic Burette, Bottletop Dispenser; 50 mL

Hirschmann | Mfr: 9392050| Item: 2550325

Operates on solar power – no more charging or changing batteries

₹396,000.00INR / Each

for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹396,000.00INR / Each

Buy It Together

Current Item

Hirschmann 9392050 Solarus Electronic Burette, Bottletop Dispenser; 50 mL
Hirschmann 9392050 Solarus Electronic Burette, Bottletop Dispenser; 50 mL
2550325

₹396,000.00INR / Each
+ +
Hirschmann 9564005 RS-232 Interface Connection Cable
2550328

₹42,000.00INR / Each
+ =
Total Price 811500.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • High accuracy
  • Burette (buret) head rotates 360° for better viewing
  • Manipulate and report data easily with RS-232 output
  • Designed with a 50 mL capacity
  • 1 year warranty

More About this Item

These digital burettes (burets) are designed for titrating in water treatment plants, material testing facilities and analytical laboratories. Integral solar panels eliminate the need for charging or changing batteries and allow for enhanced portability even in limited-lighting conditions. Easy-to-use recirculating system quickly primes the burette reducing loss of valuable reagents. Features a large LCD, user-friendly keypad, Quick-Cal individual calibration and indicator arrows to provide filling and titration direction. The hand wheels are equipped with a soft touch surface and smooth opertaing gear ensuring precise delivery of your solution. The RS-232 interface allows you to download data to your computer. RS-232 port included for computer interface. Interface cable sold separately. Valves are easily dismantled and exchangeable with provided wrench. Note: Not for use with hydrofluoric acid. Autoclavable to 250°F (121°C). What's included:recirculating tube, suction tube, thread adapters, and individual test certificate

1 Year Warranty
Specifications
  • Capacity (mL)50
  • Tolerance0.2
  • DescriptionSolarus Electronic Burette, Bottletop Dispenser; 50 mL
  • Warranty1 year
Reviews
Q&A