High-Molecular Weight HDPE Bags, Clear, 33 gal; 500/Pk

Representative image only.

High-Molecular Weight HDPE Bags, Clear, 33 gal; 500/Pk

| Item: 6010406

₹21,300.00INR / Pkg of 500

₹21,300.00INR / Pkg of 500
Product Description

More About this Item

These high-molecular weight HDPE bags have excellent impact and tensile strength. Is FDA compliant for use in food contact applications. Bags 60104-00 through -16 are clear; bag 60104-18 is metallic silver.

Specifications
  • ColorClear
  • MaterialHDPE
  • Capacity (Gal)33
  • Length (in)33
  • Width (in)40
  • Length (cm)83.82
  • Width (cm)101.6
  • Thickness (mil)0.43
  • DescriptionHigh-Molecular Weight HDPE Bags, Clear, 33 gal; 500/Pk
Reviews
Q&A