High-Molecular Weight Bags; 60 gallon, 0.86 mil

Representative image only.

High-Molecular Weight Bags; 60 gallon, 0.86 mil

| Item: 6010418

₹28,800.00INR / Case of 150

₹28,800.00INR / Case of 150
Product Description

More About this Item

These high-molecular weight HDPE bags have excellent impact and tensile strength. Is FDA compliant for use in food contact applications. Bags 60104-00 through -16 are clear; bag 60104-18 is metallic silver.

Specifications
  • ColorMetallic silver
  • MaterialHDPE
  • Capacity (Gal)60
  • Length (in)38
  • Width (in)60
  • Length (cm)96.52
  • Width (cm)152.4
  • Thickness (mil)0.86
  • DescriptionHigh-Molecular Weight Bags; 60 gallon, 0.86 mil
Reviews
Q&A