HF Scientific Online Chlorine Monitor, 100 to 240 VAC, 47 to 63 Hz

Representative image only.

HF Scientific Online Chlorine Monitor, 100 to 240 VAC, 47 to 63 Hz

HF Scientific | Mfr: 28002| Item: 9967300

Measures free or total chlorine using EPA-approved colorimetric DPD method

Call For Price

Call For Price

Buy It Together

Current Item

HF Scientific Online Chlorine Monitor, 100 to 240 VAC, 47 to 63 Hz
HF Scientific Online Chlorine Monitor, 100 to 240 VAC, 47 to 63 Hz
9967300

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description

Includes

Monitor only, order reagents, supply and drain tubing, and power cords separately.

Key Features

  • User-selectable cycle times
  • Two user-selectable alarms
  • 4 to 20 mA and RS-485 with Modbus output
  • 1 year warranty

More About this Item

The monitor uses economical and reliable colorimetric DPD (N,N-diethyl-p-phenylenediamine) chemistry to measure either free or total chlorine up to 10 mg/L. Measurement cycles can be set between 90 seconds up to 10 minutes for reagent and control optimization. Measurements are preceded by a zero measurement to compensate for color of the sample. The monitor features a scaleable 4 to 20 mA output and RS-485 with Modbus, as well as two user-selectable alarm relays.

The monitor is housed in a NEMA 4X (IP66) corrosion-resistant enclosure that comes ready to surface mount. The enclosure has a transparent cover that allow direct view of the measured value as well as the sample chamber and removable cuvette.

Sample to the unit is provided by user-supplied 1/2" OD semi-rigid or rigid tubing that connects by a push-to-connect fitting at pressures of 1 to 200 psi. The sample drain is a 1/2" hose barbed fitting; order drain hose separately. The monitor accepts 100 to 240 VAC, 47 to 63 Hz power input,;order power cord separately below or directly hard wire. Monitor does not include dry chemical reagents; order separately.

Canadian Standards Association1 Year WarrantyEPA United States Environmental protection agencyETL ListedIP 66 RatedUL Listed.
Specifications
  • Control TypeOn/Off
  • Display TypeLCD
  • Output Current4-20mA
  • Width (cm)32.7
  • Height (cm)33.34
  • Width (in)12 7/8
  • Height (in)13 1/8
  • Depth (in)5 1/16
  • DescriptionOnline Chlorine Monitor, 100 to 240 VAC, 47 to 63 Hz
  • Warranty1 year
Reviews
Q&A