HEMCO Emergency Shower/Decontamination Booth, 40" W

Representative image only.

HEMCO Emergency Shower/Decontamination Booth, 40" W

HEMCO | Mfr: 16601| Item: 3370667

Immediately rinse away harmful contaminants safely within the lab

Call For Price

Call For Price
Product Description

Includes

Frosted vinyl strip curtain, grab bars, fiberglass floor grate, and safety markings.

Key Features

  • Frosted vinyl strip curtain provides easy access and contains contaminated water splash
  • Stainless steel grab bars safely support victim and rescue personnel during decontamination
  • One-piece seamless fiberglass construction with coved corners makes cleanup easy
  • Raised flow-through floor grate prevents slippage
  • 1 year warranty

More About this Item

The emergency shower/decontamination booth quickly removes hazardous contaminates safely from an individual’s body and eyes while protecting the laboratory from water and chemical damage. The booth is equipped with an overhead drench shower and rear wall-mounted eyewash.
The shower features an immediate response activation handle, a 10” diameter plastic head and 1” IPS brass “stay-open” valve. Once activated, the shower provides a continuous stream of water for up to 15 minutes. The shower is constructed of one-piece seamless chemical-resistant fiberglass.
The eyewash includes a yellow ABS plastic bowl with two large plastic heads, and a push plate operated "stay-open" ball valve.
The fully assembled booth is ready for installation to a water supply and waste system. The 2” diameter drain outlet can be located on either the bottom or rear of unit.
Compliance: ANSI Z358.1-2009 and OSHA requirements

OSHA and ANSI recommend the emergency shower and eyewash be located within 10 seconds walking time from the hazardous site location.

1 Year WarrantyISO9001 Certified Supplier
Specifications
  • Eyewash Flow Rate (GPM)8
  • Shower Flow Rate (GPM)30
  • Inlet Connection1 1/4" IPS
  • Width (in)40
  • Depth (in)36
  • Height (in)90
  • Width (cm)101.6
  • Depth (cm)91.4
  • Height (cm)228.6
  • DescriptionCombination Shower and Eyewash Booth
  • Warranty1 year
Reviews
Q&A