Hamilton Syringe Needle Cleaning Kit with Cleaning Solution and Wires
Representative image only.
Hamilton Syringe Needle Cleaning Kit with Cleaning Solution and Wires
Hamilton | Mfr: 76620A| Item: 0793874
Removes residue—keeps needles clean
₹28,600.00INR / Each
₹28,600.00INR / Each
Product Description
Key Features
  • Reduce contamination and remove stubborn residue
  • 18 month warranty
More About this Item
The kit features 70 mL of biodegradable cleaning concentratate which helps keep needles clean by removing stubborn residues. Also included is a selection of tungsten wires in various diameters for use in cleaning plugged needles.
Not suitable for medical use. For laboratory use only.
18 Month Warranty
Specifications
  • DescriptionNeedle cleaning kit; with 70 mL of cleaning solution
  • Warranty18 months