Hamilton Syringe Guide for 25 to 500 µL Syringes
Representative image only.
Hamilton Syringe Guide for 25 to 500 µL Syringes
Hamilton | Mfr: 14906| Item: 0793876
Protect and support syringe plunger
₹12,700.00INR / Each
₹12,700.00INR / Each
Product Description
Key Features
  • Two guide arms protect the plunger from damage
  • 1 year warranty
More About this Item
Syringe guides prevent the plunger from bending or pulling out of your small-bore syringe.
1 Year Warranty
Specifications
  • Syringe TypeSyringe Guide
  • Compatible With25 to 500 µL syringes
  • DescriptionSyringe Guide for 25 to 500 µL Syringes