Hamilton Syringe Guide for 10 µL Syringes
Representative image only.
Hamilton Syringe Guide for 10 µL Syringes
Hamilton | Mfr: 14806| Item: 0793875
Protect and support syringe plunger
₹7,100.00INR / Each
₹7,100.00INR / Each
Product Description
Key Features
  • Two guide arms protect the plunger from damage
  • 1 year warranty
More About this Item
Syringe guides prevent the plunger from bending or pulling out of your small-bore syringe.
1 Year Warranty
Specifications
  • Syringe TypeSyringe Guide
  • Compatible With10 µL syringes
  • DescriptionSyringe Guide for 10 µL Syringes