Hamilton PRP-X100 Anion Exchange Column, Analytical, 250x4.1mm ID; 10um
Representative image only.
Hamilton PRP-X100 Anion Exchange Column, Analytical, 250x4.1mm ID; 10um
Hamilton | Mfr: 79433| Item: 0194204
Efficient separations with durable media
₹208,000.00INR / Each
₹208,000.00INR / Each
Product Description
Key Features
  • High selectivity, efficiency, and capacity
  • 18 month warranty
More About this Item
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene) stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-X100 Strong Anion Exchange Columns contain poly styrene-divinylbenzene)-trimethyl ammonium. These columns offer superior, rapid resolution. Exchange capacity is 0.2 mEq/g.
18 Month Warranty
Specifications
  • PhasePolystyrenedivinylbenzene
  • Column Length (mm)250
  • Column Inside Diameter (ID) (mm)4.1
  • Column Particle Size (µm)10
  • DescriptionPRP-X100 Anion Exchange Column, Analytical, 250x4.1mm ID; 10um
  • Warranty18 months