Hamilton Microliter Low-Volume Syringe Repair Kit, for 5 µL Syringe
Representative image only.
Hamilton Microliter Low-Volume Syringe Repair Kit, for 5 µL Syringe
Hamilton | Mfr: 17193| Item: 0793868
Keep your syringes working and in ideal condition
₹31,600.00INR / Each
₹31,600.00INR / Each
Product Description
Includes
needle, plunger wire, and ferrule.
Key Features
  • Repair syringes to keep your applications performing optimally
  • Compatible with low-volume syringe 07938-63
  • 1 year warranty
More About this Item
Choose repair kits suitable for your syringe. Repair kits include a needle, plunger wire, and ferrule.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • Syringe TypeRemovable Needle
  • Syringe Capacity (µL)5
  • Needle Tip Style90⁰ Blunt Tip
  • Needle IncludedYes
  • Needle Tip Number3
  • Needle Gauge24
  • Compatible With07938-63
  • DescriptionMicroliter Low-Volume Syringe Repair Kit, for 5 µL Syringe