H-B Instrument B60570-0200 Enviro-Safe Liquid-In-Glass Pocket Thermometer; -5 to 50C, Window Plastic Case, Environmentally Friendly

Representative image only.

H-B Instrument B60570-0200 Enviro-Safe Liquid-In-Glass Pocket Thermometer; -5 to 50C, Window Plastic Case, Environmentally Friendly

H-B Instrument | Mfr: B605700200| Item: 0800889

Rugged case with an eyelet at the top for easy suspension

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

H-B Instrument B60570-0200 Enviro-Safe Liquid-In-Glass Pocket Thermometer; -5 to 50C, Window Plastic Case, Environmentally Friendly
H-B Instrument B60570-0200 Enviro-Safe Liquid-In-Glass Pocket Thermometer; -5 to 50C, Window Plastic Case, Environmentally Friendly
0800889

Call For Price
+ =
Total Price 33100.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
  • 5 year warranty

More About this Item

The case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation (except closed case styles). Available in three case styles; closed metal, open metal, or closed plastic. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.

5 Year Warranty
Specifications
  • Length (")6 19/64
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Accuracy±0.5°C
  • Division (° C)0.5
  • Thermometer FillGreen Spirit
  • Overall Length (InchShort)6.3
  • Overall Length (mm)160
  • Immersion Depth (mm)160
  • Stem MaterialWhite-Back Glass
  • DescriptionEnviro-Safe Liquid-In-Glass Pocket Thermometer; -5 to 50C, Window Plastic Case, Environmentally Friendly
  • Warranty5 years
Reviews
Q&A