H-B Instrument B60302-0000 Easy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 260C, 76mm Immersion, Environmentally Friendly

Representative image only.

H-B Instrument B60302-0000 Easy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 260C, 76mm Immersion, Environmentally Friendly

H-B Instrument | Mfr: B603020000| Item: 0800843

Designed to be very easy to read

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

H-B Instrument B60302-0000 Easy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 260C, 76mm Immersion, Environmentally Friendly
H-B Instrument B60302-0000 Easy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 260C, 76mm Immersion, Environmentally Friendly
0800843

Call For Price
+ =
Total Price 33100.00 ADD 2 ITEMS TO CART
Product Description

Includes

Ring top anti-roll fitting, and NIST / DKD - traceable statement of accuracy

Key Features

  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green"
  • Statement of accuracy included
  • 5 year warranty

More About this Item

These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications where the use of mercury or other hazardous liquids is not desired. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, statement of accuracy, recyclable packaging, and are RoHS compliant.

5 Year Warranty
Specifications
  • Length (")14
  • Min Temperature (° C)-10
  • Max Temperature (° C)260
  • Accuracy±1.0°C; ±1.5°C above 100°C; ±2.5°C above 200°C; ±3.0°C above 230°C
  • Division (° C)1
  • Thermometer FillBlack Spirit
  • Overall Length (InchShort)14
  • Overall Length (mm)350
  • Immersion Depth (mm)76
  • Stem MaterialYellow-Back Glass
  • DescriptionEasy-Read General Purpose Liquid-In-Glass Thermometer; -10 to 260C, 76mm Immersion, Environmentally Friendly
  • Warranty5 years
Reviews
Q&A