GVS Total Count Swab Kit for Enumeration of all Aerobic Bacteria on Surfaces; 30/PK
Representative image only.
GVS Total Count Swab Kit for Enumeration of all Aerobic Bacteria on Surfaces; 30/PK
| Mfr: 10498315| Item: 7890946
About 1000 times more sensitive than the conventional ATP method at microorganism detection
₹92,500.00INR / Pkg of 30
₹92,500.00INR / Pkg of 30
Buy It Together
Current Item
GVS Total Count Swab Kit for Enumeration of all Aerobic Bacteria on Surfaces; 30/PK
GVS Total Count Swab Kit for Enumeration of all Aerobic Bacteria on Surfaces; 30/PK
7890946
₹92,500.00INR / Pkg of 30
+
Rapid Surface Hygiene Kit, 50 tests/kit
1404030
Call For Price
=
Total Price 92500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Capped vials with a swab, culture medium, nitrocellulose membrane device, and a certificate of conformance (CoC) with item number, lot number, and expiration date.
Key Features
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
  • For the detection of all aerobic bacteria on surfaces
  • 30/PK
  • 1 year warranty
More About this Item
Use the SwabCheck sterile cellulose swabs to quickly and easily check for the presence of bacteria on a surface. Simply wipe a surface with a swab, insert the swab into a tube containing a special medium with an indicator dye for a specific microorganism, then incubate. The super-sensitive SwabCheck medium changes color even when only a single bacterium is present. The quicker the color change happens, the higher the number of bacteria that are present. No color change after the maximum incubation period indicates the absence of the corresponding microorganism.
Use this kit for nonselective development of all aerobic bacteria on surfaces in accordance with Hazard Analysis and Critical Control Points (HACCP). Each self-standing, round-bottomed plastic vial contains yellow media with a cotton swab secured inside the screw cap. Obtain a quantitative result using the swabs, culture medium, and a membrane device included in the kit. Any growth on the membrane device with Tryptone Glucose Extract (TGE) medium confirms presence of bacteria. Store kits in temperatures between 2 and 8°C (35.6 and 46.4°F).
1 Year WarrantySterile
Specifications
  • Test TypeTotal count
  • Number of Tests Included30
  • DescriptionTotal Count Swab Kit for Enumeration of all Aerobic Bacteria on Surfaces; 30/PK