GVS Rapid Screening Hygiene SwabCheck™ Swabs; 25/PK
Representative image only.
GVS Rapid Screening Hygiene SwabCheck™ Swabs; 25/PK
| Mfr: 10498407| Item: 7890949
About 1000 times more sensitive than the conventional ATP method at microorganism detection
₹99,200.00INR / Pkg of 25
₹99,200.00INR / Pkg of 25
Buy It Together
Current Item
GVS Rapid Screening Hygiene SwabCheck™ Swabs; 25/PK
GVS Rapid Screening Hygiene SwabCheck™ Swabs; 25/PK
7890949
₹99,200.00INR / Pkg of 25
+
Rapid Surface Hygiene Kit, 50 tests/kit
1404030
Call For Price
=
Total Price 99200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Capped swabs, tubes containing a red gel media, and a certificate of conformance (CoC) with item number, lot number, and expiration date.
Key Features
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
  • For the detection of high levels of microorganisms
  • Get results in 12 hours or less
  • 25/PK
  • 1 year warranty
More About this Item
Use the SwabCheck sterile cellulose swabs to quickly and easily check for the presence of bacteria on a surface. Simply wipe a surface with a swab, insert the swab into a tube containing a special medium with an indicator dye for a specific microorganism, then incubate. The super-sensitive SwabCheck medium changes color even when only a single bacterium is present. The quicker the color change happens, the higher the number of bacteria that are present. No color change after the maximum incubation period indicates the absence of the corresponding microorganism.
The hygiene SwabCheck swabs help monitor the levels of contamination at work areas and production lines to ensure they meet the requirements for cleanliness. This rapid, same-day hygiene test changes color from red to yellow when gross bacterial and fungal contamination is detected; faster the change, higher the concentration. Use the hygiene SwabCheck swabs in conjunction with known specification levels for a more accurate assessment. Store swabs and gel media in temperatures between 5 and 25°C (41 and 77°F).
1 Year WarrantySterile
Specifications
  • Test TypeHygiene
  • Number of Tests Included25
  • DescriptionRapid Screening Hygiene SwabCheck™ Swabs; 25/PK