GVS Neutralizing Buffer Swabs, 4mL; 125/PK

Representative image only.

GVS Neutralizing Buffer Swabs, 4mL; 125/PK

| Mfr: 10498303| Item: 7890940

Provides save transfer of swabbed organisms

₹156,000.00INR / Pkg of 125

₹156,000.00INR / Pkg of 125
Product Description

Key Features

  • Qualitative and semi-quantitative hygiene control
  • Rapid results
  • Long shelf-life
  • Sterile packed and ready-for-use
  • Individually wrapped
  • 1 year warranty

More About this Item

The neutralizing buffer swabs allow you safe transfer of swabbed organisms to the laboratory without loss in viability. The buffer inactivates the bactericidal and bacteriostatic effects of chlorine and quaternary ammonium detergents. It exhibits no toxic effects on microorganisms.

Ideal for used in the monitoring of flat or convoluted surfaces for total bacterial count.

1 Year WarrantySterile
Specifications
  • MaterialCotton swab
  • Swab Total Length (InchShort)3 5/64
  • Swab Total Length (cm)7.8
  • DescriptionNeutralizing Buffer Swabs, 4mL; 125/PK
  • Warranty1 year
Reviews
Q&A