GVS Listeria SwabCheck™ Swabs; 25/PK
Representative image only.
GVS Listeria SwabCheck™ Swabs; 25/PK
| Mfr: 10498408| Item: 7890950
About 1000 times more sensitive than the conventional ATP method at microorganism detection
₹99,200.00INR / Pkg of 25
₹99,200.00INR / Pkg of 25
Buy It Together
Current Item
GVS <i>Listeria</i> SwabCheck™ Swabs; 25/PK
GVS Listeria SwabCheck™ Swabs; 25/PK
7890950
₹99,200.00INR / Pkg of 25
+
Rapid Surface Hygiene Kit, 50 tests/kit
1404030
Call For Price
=
Total Price 99200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Capped vials with media and a swab and a certificate of conformance (CoC) with item number, lot number, and expiration date.
Key Features
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
  • For the detection of Listeria on surfaces
  • 25/PK
  • 1 year warranty
More About this Item
Use the SwabCheck sterile cellulose swabs to quickly and easily check for the presence of bacteria on a surface. Simply wipe a surface with a swab, insert the swab into a tube containing a special medium with an indicator dye for a specific microorganism, then incubate. The super-sensitive SwabCheck medium changes color even when only a single bacterium is present. The quicker the color change happens, the higher the number of bacteria that are present. No color change after the maximum incubation period indicates the absence of the corresponding microorganism.
The Listeria isolation SwabCheck swabs help detect the presence of Listeria on foods and in the environment. A color change from light brown to black/dark brown indicates bacterial contamination; faster the change, higher the concentration. Each self-standing, round-bottomed plastic vial contains media with a cotton swab secured inside the screw cap. Store vials in temperatures between 2 and 8°C (35.6 and 46.4°F).
1 Year WarrantySterile
Specifications
  • Test TypeListeria
  • Number of Tests Included25
  • Genus DetectedListeria
  • DescriptionListeria SwabCheck™ Swabs; 25/PK