GVS Coliform SwabCheck™ Swabs; 25/PK
Representative image only.
GVS Coliform SwabCheck™ Swabs; 25/PK
| Mfr: 10498406| Item: 7890948
About 1000 times more sensitive than the conventional ATP method at microorganism detection
₹99,200.00INR / Pkg of 25
₹99,200.00INR / Pkg of 25
Buy It Together
Current Item
GVS Coliform SwabCheck™ Swabs; 25/PK
GVS Coliform SwabCheck™ Swabs; 25/PK
7890948
₹99,200.00INR / Pkg of 25
+
Rapid Surface Hygiene Kit, 50 tests/kit
1404030
Call For Price
=
Total Price 99200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Capped swabs, tubes containing a purple gel media, and a certificate of conformance (CoC) with item number, lot number, and expiration date.
Key Features
  • Mess-free microorganism detection
  • Offers both qualitative and semi-quantitative hygiene control
  • Sterile packed and ready to use
  • For the detection of coliforms and E. coli in the environment
  • 25/PK
  • 1 year warranty
More About this Item
Use the SwabCheck sterile cellulose swabs to quickly and easily check for the presence of bacteria on a surface. Simply wipe a surface with a swab, insert the swab into a tube containing a special medium with an indicator dye for a specific microorganism, then incubate. The super-sensitive SwabCheck medium changes color even when only a single bacterium is present. The quicker the color change happens, the higher the number of bacteria that are present. No color change after the maximum incubation period indicates the absence of the corresponding microorganism.
The coliform SwabCheck swabs provide a simple method for detecting coliforms and E. coli in the environment. Presence of these organisms indicate poor hygiene in the production process or pollution of water at source. The medium changes color from purple to yellow when coliform is present; faster color change indicates higher level of coliform. Store swabs and gel media in temperatures between 5 and 25°C (41 and 77°F).
1 Year WarrantySterile
Specifications
  • Test TypeColiform
  • Number of Tests Included25
  • DescriptionColiform SwabCheck™ Swabs; 25/PK