GVS Buffer Swabs, 4 mL; 500/PK

Representative image only.

GVS Buffer Swabs, 4 mL; 500/PK

| Mfr: 10498306| Item: 7890943

Easily collect microbial surface samples for analysis

₹476,000.00INR / Pkg of 500

₹476,000.00INR / Pkg of 500
Product Description

Includes

Capped vials with a swab and a certificate of conformance (CoC) with item number, lot number, and expiration date.

Key Features

  • Excellent for flat or convoluted surfaces
  • Does not contain bacteriostatic or bactericidal compounds
  • Sterile packed and ready-for-use
  • 500/PK
  • 1 year warranty

More About this Item

Use these cotton swabs for collecting surface contamination from flat or convoluted surfaces for analysis. Swabs do not contain any bactericidal compounds ensuring accurate results. After collecting the samples, simply transport to a laboratory for culture and enumeration. Each self-standing, round-bottomed plastic vial contains clear buffer with a cotton swab secured inside the screw cap. Store vials in temperatures between 2 and 8°C (35.6 and 46.4°F).

1 Year WarrantySterile
Specifications
  • MaterialCotton swab, polypropylene tube, and polypropylene cap with silicone washer
  • ColorWhite swab, clear tube, and blue cap
  • Swab Total Length (InchShort)3 5/64
  • Swab Total Length (cm)7.8
  • DescriptionBuffer Swabs, 4 mL; 500/PK
  • Warranty1 year
Documents & Certificates
Reviews
Q&A