Guardian Equipment G1848LH-L AutoFlow Wall-Mount Eyewash, Chrome-plated brass
Representative image only.
Guardian Equipment G1848LH-L AutoFlow Wall-Mount Eyewash, Chrome-plated brass
Guardian Equipment | Mfr: G1848LH-L| Item: 0676711
Simply pull down the sprayhead to activate flow
₹156,000.00INR / Each
₹156,000.00INR / Each
Product Description
Includes
inline strainer and ANSI compliant identification sign.
Key Features
  • Inline strainer protects sprayheads from debris in the water line
  • 2 year warranty
More About this Item
Install this wall-mount eyewash above your sink or install above a floor drain to prevent water accumulation. Features two GS-Plus™ sprayheads with a flip-top dust cover, internal flow control, and filter. Eyewash actuates upon pulling the assembly down and remains in operation until it is pushed back into the storage position. Water supply inlet is 1/2" NPT(F).
Compliance: ANSI Z385.1-2009
Two year warranty
Specifications
  • Eyewash Flow Rate (GPM)3.6
  • Eyewash Fixture MaterialChrome-plated brass
  • MountWall Mount
  • Inlet Connection1/2" NPT(F)
  • ComplianceANSI Z385.1-2009
  • DescriptionAutoFlow Wall-Mount Eyewash, Chrome-plated brass