Guardian Equipment Enclosed Safety Shower and Eyewash Station, Front Entry, Stainless Steel, Bottom Drain

Representative image only.

Guardian Equipment Enclosed Safety Shower and Eyewash Station, Front Entry, Stainless Steel, Bottom Drain

Guardian Equipment | Mfr: G2010| Item: 0185057

Immediately rinse away harmful contaminants within the lab

₹4,154,000.00INR / Each

₹4,154,000.00INR / Each
Product Description

Key Features

  • Enclosure is constructed of 16-gauge 304 stainless steel
  • Safety station features a 10” diameter shower head
  • Two GS-Plus™ spray heads with “flip-top” dust cover provide flow control and filter to remove impurities from water
  • Easily visible ANSI-compliant identification sign
  • Orange “modesty” curtain contains water and is cleanroom approved

More About this Item

The Guardian Equipment enclosed safety shower and eyewash station quickly removes hazardous contaminants from an individual’s body and eyes while protecting the laboratory from splashing water and chemical damage. The enclosure is equipped with an overhead drench shower and rear wall-mounted eyewash. The front entry features a 13-mil thick, orange vinyl curtain that is cleanroom approved, making this station ideal for areas that contain sensitive equipment that cannot be splashed with water.

The shower features an immediate response activation handle, a 10” diameter stainless steel head, and 1” IPS stainless steel “stay-open” valve. The three-sided enclosure is constructed of 16-gauge 304 stainless steel with #4 satin finish and welded seams. Unit is completely assembled and water tested prior to shipment.

Specifications
  • Shower Flow Rate (GPM)20
  • Eyewash Fixture MaterialStainless Steel
  • Tank MaterialStainless Steel
  • Shower Head MaterialStainless Steel
  • Supply Pipe MaterialStainless Steel
  • MountFloor
  • Outlet Connections4" NPT(F)
  • Inlet Connection1" NPT(F)
  • Width (in)33
  • Depth (in)36 5/8
  • Height (in)87 3/4
  • Width (cm)83.8
  • Depth (cm)93
  • Height (cm)222.9
  • ComplianceANSI Z385.1-2009
  • DescriptionEnclosed Safety Shower and Eyewash Station, Front Entry, Stainless Steel, Bottom Drain
Reviews
Q&A