Glo Germ 1003 Glo-Germ portable sanitation training kit
Representative image only.
Glo Germ 1003 Glo-Germ portable sanitation training kit
Glo Germ | Mfr: 1003| Item: 1410303
₹12,400.00INR / Each
₹12,400.00INR / Each
Buy It Together
Current Item
Glo Germ 1003 Glo-Germ portable sanitation training kit
Glo Germ 1003 Glo-Germ portable sanitation training kit
1410303
₹12,400.00INR / Each
+
Glo Germ GGL Replacement Oil for Training Kit, 8 oz
1410304
₹4,800.00INR / Each
=
Total Price 17200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Portable model 14103-03 includes a 6-1/2'' long, battery-operated LED UV flashlight.
Key Features
  • Ideal for training employees about proper hand washing and cleaning techniques
More About this Item
Kits contain lotion and powder which contain plastic ''germs'' that glow under ultraviolet (UV) light. To determine proper hand washing technique , apply oils to hands. This spreads thousands of these tiny plastic flourescent ''germs'' on the hands. Place hands under the UV light after normal hand washing. Plastic germs glow brightly to show the spots missed or not cleaned properly.
Kit comes with an 8-oz bottle of oil and 2-oz bottle of powder.
Specifications
  • DescriptionGlo-Germ portable sanitation training kit