Glas-Col R34 561 Heater Block, 24 Holes, holds 15mm vials
Representative image only.
Glas-Col R34 561 Heater Block, 24 Holes, holds 15mm vials
Glas-Col | Mfr: R34 561| Item: 3649510
₹70,000.00INR / Each
₹70,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
These heating blocks are constructed of anodized aluminum and designed for optimum heat transfer to tube or vial sample contents. Select heating block based tube or vial size.
Conformite Europeenne1 Year Warranty
Specifications
  • DescriptionHeater Block, 24 Holes, holds 15mm vials