Glas-Col Cord Adapter From IEC 60320 to CEE7 (Europe Plug), 12" L; 10A, 240V
Representative image only.
Glas-Col Cord Adapter From IEC 60320 to CEE7 (Europe Plug), 12" L; 10A, 240V
Glas-Col | Mfr: R08-434| Item: 8980019
₹19,700.00INR / Each
₹19,700.00INR / Each
Product Description
Key Features
  • Europe specific female plug
  • 1 year warranty
More About this Item
Choose an interchangeable cord/receptacle assembly for your 230 VAC temperature controller. Each 12" (30.5 cm) cord set features a country-specific female plug on one end and an IEC 60320 C19 male plug on the other end rated for 10 A at 240 VAC.
1 Year Warranty
Specifications
  • Power (VAC)240
  • DescriptionCord Adapter From IEC 60320 to CEE7 (Europe Plug), 12" L; 10A, 240V