Glas-Col 100E O540 Support for Flexible Fabric Heating Mantle, 2000 Ml
Representative image only.
Glas-Col 100E O540 Support for Flexible Fabric Heating Mantle, 2000 Ml
Glas-Col | Mfr: 100E O540| Item: 3622585
Safely support flasks and mantles
₹33,100.00INR / Each
₹33,100.00INR / Each
Product Description
Key Features
  • Free up valuable bench space while heating solutions
  • 1 year warranty
More About this Item
Use these extension supports for attaching hemispherical and spherical mantle to lattice. Each extension is made of a steel ring and heat resistant strap basket.
See our clamps and supports section for support rods and frames.
1 Year Warranty
Specifications
  • Max Flask Size (mL)2000
  • MaterialSteel
  • DescriptionSupport for Flexible Fabric Heating Mantle, 2000 Ml