GFS Chemicals Turbidity Standards Kit for HF Scientific Micro TPW meter, 0, 10, 1000 NTU
Representative image only.
GFS Chemicals Turbidity Standards Kit for HF Scientific Micro TPW meter, 0, 10, 1000 NTU
GFS Chemicals | Mfr: 86226| Item: 0839170
Ensure accurate readings by calibrating your turbidimeter
₹81,200.00INR / Each
₹81,200.00INR / Each
Buy It Together
Current Item
GFS Chemicals Turbidity Standards Kit for HF Scientific Micro TPW meter, 0, 10, 1000 NTU
GFS Chemicals Turbidity Standards Kit for HF Scientific Micro TPW meter, 0, 10, 1000 NTU
0839170
₹81,200.00INR / Each
+ =
Total Price 128800.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Designed especially for your Cole-Parmer®, Hach, LaMotte®, or HF turbidimeter
More About this Item
Kits include a 250-mL bottle of primary standard, a secondary standard-filled cuvette for each of the NTU values listed, and two empty glass cuvettes. All cuvettes are designed to fit your brand-name meter.
Specifications
  • NTU Value0, 10, 1000
  • Volume (mL)250
  • Volume (oz)8.5
  • Use withHF Scientific Micro TPW Turbidimeter
  • DescriptionTurbidity Standards Kit for HF Scientific Micro TPW meter, 0, 10, 1000 NTU