GFS Chemicals Turbidity Standards Kit for HF Scientific Micro 100 Meters, 0, 10, 1000 NTU
Representative image only.
GFS Chemicals Turbidity Standards Kit for HF Scientific Micro 100 Meters, 0, 10, 1000 NTU
GFS Chemicals | Mfr: 85555| Item: 0839129
Ensure accurate readings by calibrating your turbidimeter
₹84,900.00INR / Each
₹84,900.00INR / Each
Buy It Together
Current Item
GFS Chemicals Turbidity Standards Kit for HF Scientific Micro 100 Meters, 0, 10, 1000 NTU
GFS Chemicals Turbidity Standards Kit for HF Scientific Micro 100 Meters, 0, 10, 1000 NTU
0839129
₹84,900.00INR / Each
+ =
Total Price 132500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Designed especially for your Cole-Parmer®, Hach, LaMotte®, or HF turbidimeter
More About this Item
Kits include a 250-mL bottle of primary standard, a secondary standard-filled cuvette for each of the NTU values listed, and two empty glass cuvettes. All cuvettes are designed to fit your brand-name meter.
Specifications
  • Conductivity Value (µS)1413
  • Resistivity Value (MΩ)0.00071
  • NTU Value0, 10, 1000
  • Volume (mL)250
  • Volume (oz)8.5
  • Use withHF Scientific Micro 100 Turbidimeter
  • DescriptionTurbidity Standards Kit for HF Scientific Micro 100 Meters, 0, 10, 1000 NTU