GFS Chemicals Turbidity Standards Kit for Hach 2100 N/AN Meters; 0/20/200/1000/4000 NTU

Representative image only.

GFS Chemicals Turbidity Standards Kit for Hach 2100 N/AN Meters; 0/20/200/1000/4000 NTU

GFS Chemicals | Mfr: 85551| Item: 0839124

Ensure accurate readings by calibrating your turbidimeter

₹144,000.00INR / Each

₹144,000.00INR / Each

Buy It Together

Current Item

GFS Chemicals Turbidity Standards Kit for Hach 2100 N/AN Meters; 0/20/200/1000/4000 NTU
GFS Chemicals Turbidity Standards Kit for Hach 2100 N/AN Meters; 0/20/200/1000/4000 NTU
0839124

₹144,000.00INR / Each
+ =
Total Price 191600.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Designed especially for your Cole-Parmer®, Hach, LaMotte®, or HF turbidimeter

More About this Item

Kits include a 250-mL bottle of primary standard, a secondary standard-filled cuvette for each of the NTU values listed, and two empty glass cuvettes. All cuvettes are designed to fit your brand-name meter.

Specifications
  • NTU Value0, 20, 200, 1000, 4000
  • Volume (mL)250
  • Volume (oz)8.5
  • Use withHach 2100 N/AN Laboratory Turbidimeters
  • DescriptionTurbidity Standards Kit for Hach 2100 N/AN Meters; 0/20/200/1000/4000 NTU
Reviews
Q&A