GFS Chemicals Turbidity Standards Kit for and ICM meters, 0.5, 5.0, 40 NTU
Representative image only.
GFS Chemicals Turbidity Standards Kit for and ICM meters, 0.5, 5.0, 40 NTU
GFS Chemicals | Mfr: 85543| Item: 0839114
Ensure accurate readings by calibrating your turbidimeter
₹129,000.00INR / Each
₹129,000.00INR / Each
Buy It Together
Current Item
GFS Chemicals Turbidity Standards Kit for and ICM meters, 0.5, 5.0, 40 NTU
GFS Chemicals Turbidity Standards Kit for and ICM meters, 0.5, 5.0, 40 NTU
0839114
₹129,000.00INR / Each
+ =
Total Price 176600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Designed especially for your Cole-Parmer®, Hach, LaMotte®, or HF turbidimeter
More About this Item
Kits include a 250-mL bottle of primary standard, a secondary standard-filled cuvette for each of the NTU values listed, and two empty glass cuvettes. All cuvettes are designed to fit your brand-name meter.
Specifications
  • NTU Value0.5, 5.0, 40
  • Volume (mL)250
  • Volume (oz)8.5
  • Use withCole-Parmer EPA-Recognized Turbidimeters
  • DescriptionTurbidity Standards Kit for and ICM meters, 0.5, 5.0, 40 NTU