GF Signet 3-8050-1 Universal Mount Junction Box
Representative image only.
GF Signet 3-8050-1 Universal Mount Junction Box
GF Signet | Mfr: 3-8050-1| Item: 6847665
Junction boxes provide convenient wiring
₹21,400.00INR / Each
₹21,400.00INR / Each
Product Description
Includes
Top cover, transmitter base, universal mounting plate and bracket, liquid tight connector kit.
Key Features
  • Mount on a wall, pipe, or tank
  • Two universal terminal blocks that enable cable extensions for pH, ORP, flow, temperature, pressure, and conductivity electrodes
  • 2 year warranty
More About this Item
Simplify wiring and enable cable extension with junction boxes.
Two year warranty
Specifications
  • Compatible WithpH, ORP, flow, temperature, pressure, and conductivity sensors/electrodes (2750-1, 2750-3, 2750-4, 2839-2842 [-1, -1D], 2350, 2450)
  • DescriptionUniversal Mount Junction Box
  • Warranty2 Years