Generator Probe Tool Kit for HO-200 and HO-400 Series Homogenizers
Representative image only.
Generator Probe Tool Kit for HO-200 and HO-400 Series Homogenizers
| Item: 7890547
Keep your homogenizer up and running
₹22,900.00INR / Each
₹22,900.00INR / Each
Product Description
More About this Item
Ideal replacement parts for your Cole-Parmer branded homogenizers and probes.
The kit includes knife tool to remove 5, 7, and 10mm probe knives, bearing removal tools for 5 and 7mm generator probes, and a
shaft holder.
Specifications
  • DescriptionGenerator Probe Tool Kit for HO-200 and HO-400 Series Homogenizers
  • WarrantyNone