General Tools DCS400-PE Wireless Datalogging Video Inspection Camera/Borescope, 14 mm Electrically Insulated Probe, 3.5" Color LCD
Representative image only.
General Tools DCS400-PE Wireless Datalogging Video Inspection Camera/Borescope, 14 mm Electrically Insulated Probe, 3.5" Color LCD
General Tools | Mfr: DCS600A| Item: 4861056
Ideal for inspection of duct work, piping, and wiring locations
₹70,100.00INR / Each
₹70,100.00INR / Each
Product Description
Includes
Probe, MicroSD memory ( 2 GB), rechargeable Lithium-ion batteries, AC2 adaptor/battery charger, USB and A/V cables, "Y" charging cable splitter, and custom hard plastic carrying case.
Key Features
  • Flexible camera-tipped probes
  • Adjustable LED lighting
  • Captures still and video images
  • 3 year warranty
More About this Item
These video borescope/inspection scopes are ideal for inspection of duct work, piping, wiring locations, and much more. Camera-tipped probes are flexible and strong. View findings instantaneously on the LCD monitor; some models will capture still and video images. Adjustable LED lighting on the probe tip provides illumination in dimly lit areas, plus allows you to lower the light intensity when viewing reflective objects to eliminate washout.
DCS400-PE
* For the inspection of inaccessible areas with exposed electrical wiring
* ETL certified for CAT III-1000 V, CAT IV-600 V use
* 9/16" (14 mm) diameter electrically insulated flexible probe
* Monitor can be detached for remote viewing up to 32 ft (10 m) away
* View results in real time, record up to 3 hours of video and capture still photos with instant playback
3 Year WarrantyConformite Europeenne
Specifications
  • DescriptionWireless Video Inspection Scope, 3.5" color LCD, 14mm Electrically Insulated Probe
  • Warranty3 Years