Fytertech Spilfyter Kolor-Safe Base Neutralizer, 1 quart
Representative image only.
Fytertech Spilfyter Kolor-Safe Base Neutralizer, 1 quart
Fytertech | Mfr: 430001| Item: 8642312
Three-color change lets you know when spill is completely neutralized
₹5,000.00INR / Quart of 1
₹5,000.00INR / Quart of 1
Product Description
More About this Item
Protect your workers and facility from base spills by quickly and efficiently neutralizing them. Simply apply the neutralizer until the spill goes through the three-color change. Base neutralizer changes from red to blue to yellow to indicate total neutralization.
Specifications
  • DescriptionBase Neutralizer, 1 quart