Fuego Gasprofi 1 Micro Burner; Natural Gas, Propane/Butane
Representative image only.
Fuego Gasprofi 1 Micro Burner; Natural Gas, Propane/Butane
Fuego | Mfr: 6.004.000| Item: 3630201
Compact, economical, gas burner
₹206,000.00INR / Each
₹206,000.00INR / Each
Product Description
Includes
flame monitor and gas nozzles, assembly tools, tubing connector, and switching (global) power supply.
Key Features
  • Flame monitor automatically shuts gas supply off if flame extinquishes accidentally
  • Simple and safe push button operation -- no matches needed
  • Energy-saving consumption
  • 2 year warranty
More About this Item
This Gasprofi series incorporates advanced heating and flame sterilization technology. A flame monitor ensures safe operation and shuts off gas flow if the flame inadvertently extinguishes. All external components are constructed of stainless steel for maximum sterility, and the removable burner head allows thorough cleaning. Use with natural gas or propane/butane gases.
Two year warrantyConformite Europeenne
Specifications
  • Gas TypeNatural Gas, Propane/Butane
  • DescriptionMicro Burner; Natural Gas, Propane/Butane
  • Warranty2 Years