Fuego 8.001.000 Scs Basic Laboratory Gas Burner; Natural Gas, Propane/Butane

Representative image only.

Fuego 8.001.000 Scs Basic Laboratory Gas Burner; Natural Gas, Propane/Butane

Fuego | Mfr: 8,201,000| Item: 3630202

Simple and safe operation for all flame related applications in the laboratory

₹171,000.00INR / Each

₹171,000.00INR / Each

Buy It Together

Current Item

Fuego 8.001.000 Scs Basic Laboratory Gas Burner; Natural Gas, Propane/Butane
Fuego 8.001.000 Scs Basic Laboratory Gas Burner; Natural Gas, Propane/Butane
3630202

₹171,000.00INR / Each
+
Fuego 6.000.501 Global Switching Power Supply
3630210

₹11,300.00INR / Each
=
Total Price 182300.00 ADD 2 ITEMS TO CART
Product Description

Includes

global power supply adapter (100 to 240 VAC, 50/60 Hz), tilt mechanism, holding device for 3 inoculation loop holder and nozzles for natural gas, propane, and butane

Key Features

  • Ideal for use in all industrial, laboratory, and cleanroom settings
  • Modern low-profile body design reduces air flow disturbances and is very easy to clean
  • Robust, all stainless steel construction and fireproof controls
  • 2 year warranty

More About this Item

With an ergonomic design and extensive standard features, these laboratory gas burners withstand extreme lab conditions while incorporating innovative safety technology. The Safety Control System (SCS) constantly analyzes potential hazards, and, if necessary, initiates safety measures, such as an interruption of the gas supply. An adjustable automatic cut-off system inhibits unintentional ignition. To protect you from burns after prolonged use, the residual heat display will signal that the burner head is still hot. Burners also feature continuous burner head control which identifies burner head clogging and ensures correct assembly of burner head.

Fuego SCS Basic Burner can be operated with a foot pedal or button function. Three programs are available: a
flexible start/stop function with 60-minute timer and the conventional foot-pedal operation for short flame sterilization. Additionally the flame can be started and stopped by pushing the function knob.(Foot pedal included.)

UL Listed to Canadian safety standards.Two year warranty
Specifications
  • Gas TypeNatural Gas, Propane/Butane
  • Inlet Tube OD (in)0.25
  • Width (in)5 1/2
  • Length (in)4
  • Height (in)1 15/16
  • Power (VAC)120 / 240
  • DescriptionScs Basic Laboratory Gas Burner; Natural Gas, Propane/Butane
  • Warranty2 Years
Reviews
Q&A