Frigimat Junior Dry Ice Maker; Collection Bag, Clamp, and Plastic Cone
Representative image only.
Frigimat Junior Dry Ice Maker; Collection Bag, Clamp, and Plastic Cone
Frigimat | Mfr: F388860000| Item: 0371632
Produces dry ice in flake or snow
₹62,500.00INR / Each
₹62,500.00INR / Each
Buy It Together
Current Item
Frigimat Junior Dry Ice Maker; Collection Bag, Clamp, and Plastic Cone
Frigimat Junior Dry Ice Maker; Collection Bag, Clamp, and Plastic Cone
0371632
₹62,500.00INR / Each
+
Frigimat F388860001 Extra Bags for Junior Dry Ice Maker, 3/pk
0371690
₹32,900.00INR / Pkg of 3
=
Total Price 95400.00 ADD 2 ITEMS TO CART
Product Description
Includes
collection bag, clamp, and plastic cone.
Key Features
  • Produces only the dry ice you need
  • Quick and easy to use
More About this Item
Operate these dry ice makers by screwing the nozzle directly to a cylinder of compressed LCO2 with a CGA connector. Note: cylinder must have a dip tube. Bag is secured to cone on the nozzle with the clamp. Compressed LCO2 expands into the bag and creates dry-ice snow. The ice may be squeezed into a hard pack or left in flake form depending on intended use. It makes 0.77 kg (1.7 lb) of flakes per bag. The cone is 13 cm (5") long x 6 cm (2-5⁄16") diameter at end. The bag is 15 cm (6") W x 30 cm (12") L.
Specifications
  • DescriptionJunior Dry Ice Maker; Collection Bag, Clamp, and Plastic Cone