Foxx Life Sciences Versatile Carboy Cap, 83B, Open with Solid Adapter; 2/PK
Representative image only.
Foxx Life Sciences Versatile Carboy Cap, 83B, Open with Solid Adapter; 2/PK
| Mfr: 205-1004-RLS| Item: 0605924
Leakproof caps with the flexibility of interchangeable ports
₹44,300.00INR / Pkg of 2
₹44,300.00INR / Pkg of 2
Product Description
Key Features
  • Finger grips allow easy tightening and opening
  • Choose from a variety of sizes and configurations*
  • Change the top connections of each carboy to match your application needs
  • Autoclavable
More About this Item
Versatile carboy caps are made of autoclavable polypropylene and come in industry-standard sizes.
Solid caps do not accept insert adapters.
Specifications
  • Cap MaterialPP
  • Cap / Stopper Size83B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing IDOpen with Solid Adapter
  • DescriptionVersatile Carboy Cap, 83B, Open with Solid Adapter; 2/Pk