Fowler 54-225-555 Fred V Xtra-Value Depth Gauge
Representative image only.
Fowler 54-225-555 Fred V Xtra-Value Depth Gauge
Fowler | Mfr: 542255550| Item: 1050801
Call For Price
for an additional
₹14,200.00INR / Each
for an additional
₹25,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Fowler 54-225-555 Fred V Xtra-Value Depth Gauge
Fowler 54-225-555 Fred V Xtra-Value Depth Gauge
1050801
Call For Price
+
InnoCal NIST-Traceable Calibration; Height Gauge, 0-24"
1705014
₹14,200.00INR / Each
=
Total Price 14200.00 ADD 2 ITEMS TO CART
Product Description
Includes
plastic case
Key Features
  • Read in inch, metric, or fraction
  • Spring-loaded and hold measurement button for ease of use
  • Large display
  • Range: 0 to 16"/406 mm
  • Resolution: 0.0005"/0.01 mm (Fractions in 1/64" increments)
  • 1 year warranty
1 Year Warranty
Specifications
  • DescriptionFred V Xtra-Value Depth Gauge