Fowler 54-115-333 Simplex Opto Cable with RS-232 Connection
Representative image only.
Fowler 54-115-333 Simplex Opto Cable with RS-232 Connection
Fowler | Mfr: 541153330| Item: 9715250
Optimize performance of your equipment
Call For Price
Call For Price
Product Description
Includes
6-ft (1.8-m) L cable, 9-pin connector, and demo software.
Key Features
  • Make quick and easy connections for data transfer
  • 1 year warranty
More About this Item
Easily transfer your data for analysis with these connection cables.
1 Year Warranty
Specifications
  • Compatible WithDIgital caliper and micrometer set 09923-41
  • ConnectionsOpto to RS-232
  • DescriptionSimplex Opto Cable with RS-232 Connection