Fowler 52-580-015 Micrometer Mounting Stand
Representative image only.
Fowler 52-580-015 Micrometer Mounting Stand
Fowler | Mfr: 525800150| Item: 0992411
Support micrometers and other instruments for easier measurement
₹109,000.00INR / Each
₹109,000.00INR / Each
Product Description
Key Features
  • Hold your measuring device in place
  • Working height of 6.5" (16.5 cm)
  • Holds indicators with stems of 3/8" and 8 mm
  • 3-1/2" x 3-1/2" working surface
  • 1 year warranty
More About this Item
No need to hold on to your devices, these support stands firmly hold your measuring instruments for accurate measurements.
1 Year Warranty
Specifications
  • Compatible WithIndicators and probes for thickness and comparative measurements
  • DescriptionMounting Stand for Indicators and Probes